Anti-DDX5 Goat polyclonal antibody

Proveedor: OriGene
ORIGTA302658EA 547 EUR
ORIGTA302658
Anti-DDX5 Goat polyclonal antibody
Anticuerpos
Recommended Dilutions: Western Blot: 0.3-1?g/ml; Immunochemistry: 2-5?g/ml

Type: Primary
Antigen: DDX5 (DEAD (Asp-Glu-Ala-Asp) box helicase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR