Anti-APOE Goat polyclonal antibody

Proveedor: OriGene
ORIGTA302644EA 500 EUR
ORIGTA302644
Anti-APOE Goat polyclonal antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR