Anti-AMPH Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302642EA 502 EUR
ORIGTA302642
Anti-AMPH Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:16000 Western Blot: 01-05µg/ml

Type: Primary
Antigen: AMPH (amphiphysin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR