Anti-WISP1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302635EA 547 EUR
ORIGTA302635
Anti-WISP1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:128000 Western Blot: 01-03µg/ml

Type: Primary
Antigen: WISP1 (WNT1 inducible signaling pathway protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR