Anti-UCP2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302628EA 500 EUR
ORIGTA302628
Anti-UCP2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: UCP2 (uncoupling protein 2 (mitochondrial, proton carrier))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR