Anti-TCF3 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302623EA 547 EUR
ORIGTA302623
Anti-TCF3 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:128000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: TCF3 (transcription factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR