Anti-SORD Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302618EA 500 EUR
ORIGTA302618
Anti-SORD Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 01-1µg/ml

Type: Primary
Antigen: SORD (sorbitol dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Chicken, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR