Anti-SOCS3 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302617EA 500 EUR
ORIGTA302617
Anti-SOCS3 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 4-6µg/ml

Type: Primary
Antigen: SOCS3 (suppressor of cytokine signaling 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR