Anti-SNX16 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302615EA 550 EUR
ORIGTA302615
Anti-SNX16 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 005-02µg/ml

Type: Primary
Antigen: SNX16 (sorting nexin 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR