Anti-HLTF Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302613EA 547 EUR
ORIGTA302613
Anti-HLTF Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 02-1µg/ml

Type: Primary
Antigen: HLTF (helicase-like transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR