Anti-SET Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302603EA 500 EUR
ORIGTA302603
Anti-SET Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: SET (SET nuclear oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR