Anti-SKAP2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302599EA 547 EUR
ORIGTA302599
Anti-SKAP2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.5-2µg/ml.An

Type: Primary
Antigen: SKAP2 (src kinase associated phosphoprotein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR