Anti-PADI4 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302558EA 547 EUR
ORIGTA302558
Anti-PADI4 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-2µg/ml

Type: Primary
Antigen: PADI4 (peptidyl arginine deiminase, type IV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR