Anti-POU5F1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302555EA 547 EUR
ORIGTA302555
Anti-POU5F1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: POU5F1 (POU class 5 homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR