Anti-KPNA3 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302524EA 500 EUR
ORIGTA302524
Anti-KPNA3 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1/32000 Western Blot: 01-05µg/ml; Immunochemistry: 5-10µg/ml

Type: Primary
Antigen: KPNA3 (karyopherin alpha 3 (importin alpha 4))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR