Anti-IRF2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302517EA 502 EUR
ORIGTA302517
Anti-IRF2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: IRF2 (interferon regulatory factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR