Anti-IGFBP6 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302514EA 550 EUR
ORIGTA302514
Anti-IGFBP6 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: IGFBP6 (insulin-like growth factor binding protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR