Anti-FABP2 Goat polyclonal antibody

Proveedor: OriGene
ORIGTA302479EA 547 EUR
ORIGTA302479
Anti-FABP2 Goat polyclonal antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0001µg/ml

Type: Primary
Antigen: FABP2 (fatty acid binding protein 2, intestinal)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR