Anti-ELF3 Goat polyclonal antibody

Proveedor: OriGene
ORIGTA302473EA 547 EUR
ORIGTA302473
Anti-ELF3 Goat polyclonal antibody
Anticuerpos
Recommended Dilutions: Western Blot: 2-4µg/ml

Type: Primary
Antigen: ELF3 (E74-like factor 3 (ets domain transcription factor, epithelial-specific ))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR