Anti-BNIP1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302435EA 500 EUR
ORIGTA302435
Anti-BNIP1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:8000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: BNIP1 (BCL2/adenovirus E1B 19kDa interacting protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR