Anti-BDH2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302431EA 547 EUR
ORIGTA302431
Anti-BDH2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: BDH2 (3-hydroxybutyrate dehydrogenase, type 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR