Anti-SIL1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302430EA 500 EUR
ORIGTA302430
Anti-SIL1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-2µg/ml

Type: Primary
Antigen: SIL1 (SIL1 nucleotide exchange factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR