Anti-ASF1A Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302422EA 500 EUR
ORIGTA302422
Anti-ASF1A Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: ASF1A (anti-silencing function 1A histone chaperone)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR