Anti-AIP Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302405EA 547 EUR
ORIGTA302405
Anti-AIP Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 002-006µg/ml

Type: Primary
Antigen: AIP (aryl hydrocarbon receptor interacting protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR