Anti-SFN Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA302401EA 550 EUR
ORIGTA302401
Anti-SFN Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: SFN (stratifin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR