Anti-CD14 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA302271EA 638 EUR
ORIGTA302271
Anti-CD14 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR