Anti-UBTD1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA302267EA 637 EUR
ORIGTA302267
Anti-UBTD1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: UBTD1 (ubiquitin domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR