Anti-MEF2C Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA302220EA 638 EUR
ORIGTA302220
Anti-MEF2C Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:10-50; Immunochemistry: 1:10-50

Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR