Anti-PIAS1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA302213EA 698 EUR
ORIGTA302213
Anti-PIAS1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: PIAS1 (protein inhibitor of activated STAT, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR