Anti-CDH8 Rabbit polyclonal antibody

Proveedor: OriGene
ORIGTA302196EA 697 EUR
ORIGTA302196
Anti-CDH8 Rabbit polyclonal antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: CDH8 (cadherin 8, type 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR