Anti-CSNK2B Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA302170EA 638 EUR
ORIGTA302170
Anti-CSNK2B Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CSNK2B (casein kinase 2, beta polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR