Anti-PDK2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA302110EA 698 EUR
ORIGTA302110
Anti-PDK2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: PDK2 (pyruvate dehydrogenase kinase, isozyme 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR