Anti-PRMT8 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA302105EA 638 EUR
ORIGTA302105
Anti-PRMT8 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PRMT8 (protein arginine methyltransferase 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR