Anti-GFAP Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA302094EA 637 EUR
ORIGTA302094
Anti-GFAP Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-100; Western Blot: 1:1000

Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR