Anti-NR0B1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA302082EA 638 EUR
ORIGTA302082
Anti-NR0B1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: NR0B1 (nuclear receptor subfamily 0, group B, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR