Anti-SSR1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA302066EA 637 EUR
ORIGTA302066
Anti-SSR1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:10-50; Western Blot: 1:1000

Type: Primary
Antigen: SSR1 (signal sequence receptor, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR