Anti-PDK4 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA302046EA 637 EUR
ORIGTA302046
Anti-PDK4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:100; Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: PDK4 (pyruvate dehydrogenase kinase, isozyme 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR