Anti-APOE Rabbit polyclonal antibody

Proveedor: OriGene
ORIGTA302010EA 698 EUR
ORIGTA302010
Anti-APOE Rabbit polyclonal antibody
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR