Anti-STRN Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA301717EA 658 EUR
ORIGTA301717
Anti-STRN Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: STRN (striatin, calmodulin binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR