Anti-TERT Mouse Monoclonal Antibody [clone: 2D8]

Proveedor: OriGene
ORIGTA301587EA 658 EUR
ORIGTA301587
Anti-TERT Mouse Monoclonal Antibody [clone: 2D8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1: 1000

Type: Primary
Antigen: TERT (telomerase reverse transcriptase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR