Anti-CERK Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA301540EA 569 EUR
ORIGTA301540
Anti-CERK Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: CERK (ceramide kinase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR