Anti-H3F3B Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA301483EA 615 EUR
ORIGTA301483
Anti-H3F3B Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1: 1000

Type: Primary
Antigen: H3F3B (H3 histone, family 3B (H3.3B))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR