Anti-GLUL Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA301447EA 601 EUR
ORIGTA301447
Anti-GLUL Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: GLUL (glutamate-ammonia ligase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR