Anti-KDR Rabbit Monoclonal Antibody [clone: EPRER16Y]

Proveedor: OriGene
ORIGTA301154EA 755 EUR
ORIGTA301154
Anti-KDR Rabbit Monoclonal Antibody [clone: EPRER16Y]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunoprecipitation: 1:20

Type: Primary
Antigen: KDR (kinase insert domain receptor (a type III receptor tyrosine kinase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR