Anti-PPP1CB Rabbit Monoclonal Antibody [clone: EP1804Y]

Proveedor: OriGene
ORIGTA300926EA 770 EUR
ORIGTA300926
Anti-PPP1CB Rabbit Monoclonal Antibody [clone: EP1804Y]
Anticuerpos
Recommended Dilutions: Western Blot: 1:100000-1:200000; Immunoprecipitation: 1:100; Flow Cytometry: 1:25

Type: Primary
Antigen: PPP1CB (protein phosphatase 1, catalytic subunit, beta isozyme)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR