Anti-MAP2K4 Rabbit Monoclonal Antibody [clone: EP1075Y]

Proveedor: OriGene
ORIGTA300743EA 698 EUR
ORIGTA300743
Anti-MAP2K4 Rabbit Monoclonal Antibody [clone: EP1075Y]
Anticuerpos
Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: 1:50 100; Immunoprecipitation: 1:50

Type: Primary
Antigen: MAP2K4 (mitogen-activated protein kinase kinase 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR