Anti-BLNK Rabbit Monoclonal Antibody [clone: Y491]

Proveedor: OriGene
ORIGTA300524EA 624 EUR
ORIGTA300524
Anti-BLNK Rabbit Monoclonal Antibody [clone: Y491]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunoprecipitation: 1:20

Type: Primary
Antigen: BLNK (B-cell linker)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR