Anti-CASP3 Rabbit monoclonal antibody [clone: E83-103]

Proveedor: OriGene
ORIGTA300422EA 748 EUR
ORIGTA300422
Anti-CASP3 Rabbit monoclonal antibody [clone: E83-103]
Anticuerpos
Recommended Dilutions: Immunochemistry/Immunofluorescence: 5 ?g/ml; Western Blot: 1:10000; Immunoprecipitation: 1:100; Flow Cytometry: 1:50; Immunochemistry: 1:250

Type: Primary
Antigen: CASP3 (caspase 3, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR