Anti-PKM Mouse Monoclonal Antibody [clone: 3C6]

Proveedor: OriGene
ORIGTA190160EA 512 EUR
ORIGTA190160
Anti-PKM Mouse Monoclonal Antibody [clone: 3C6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: PKM (pyruvate kinase, muscle)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR