Anti-eGFP Mouse Monoclonal Antibody

Proveedor: OriGene
ORIGTA150052EA 497 EUR
ORIGTA150052
Anti-eGFP Mouse Monoclonal Antibody
Anticuerpos
Type: Tag
Antigen: eGFP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR